Merck
  • Constitutive μ-opioid receptor activity leads to long-term endogenous analgesia and dependence.
Constitutive μ-opioid receptor activity leads to long-term endogenous analgesia and dependence.
Science (New York, N.Y.) (2013-09-21)
G Corder, S Doolen, R R Donahue, M K Winter, B L Jutras, Y He, X Hu, J S Wieskopf, J S Mogil, D R Storm, Z J Wang, K E McCarson, B K Taylor
ABSTRACT
Opioid receptor antagonists increase hyperalgesia in humans and animals, which indicates that endogenous activation of opioid receptors provides relief from acute pain; however, the mechanisms of long-term opioid inhibition of pathological pain have remained elusive. We found that tissue injury produced μ-opioid receptor (MOR) constitutive activity (MOR(CA)) that repressed spinal nociceptive signaling for months. Pharmacological blockade during the posthyperalgesia state with MOR inverse agonists reinstated central pain sensitization and precipitated hallmarks of opioid withdrawal (including adenosine 3',5'-monophosphate overshoot and hyperalgesia) that required N-methyl-D-aspartate receptor activation of adenylyl cyclase type 1. Thus, MOR(CA) initiates both analgesic signaling and a compensatory opponent process that generates endogenous opioid dependence. Tonic MOR(CA) suppression of withdrawal hyperalgesia may prevent the transition from acute to chronic pain.
Product Description
Supelco
6β-Naltrexol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Naltrexone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Adenosine 2′(3′)-monophosphate mixed isomers