Merck
  • The murine common deletion: mitochondrial DNA 3,860-bp deletion after irradiation.
The murine common deletion: mitochondrial DNA 3,860-bp deletion after irradiation.
Radiation research (2013-09-26)
Steven B Zhang, David Maguire, Mei Zhang, Zhenhuan Zhang, Amy Zhang, Liangjie Yin, Lurong Zhang, Luquiang Huang, Sadasivan Vidyasagar, Steven Swarts, Paul Okunieff
ABSTRACT
This study demonstrates that mice, similar to humans, have a common mitochondrial DNA deletion (3,860 bp) that encodes 5 transfer RNA genes and 5 polypeptide genes that is related to aging, tissue type and radiotoxicity. Our research indicates that the deletion ratio in the liver was significantly higher than in the brain and gut tissues of 8-month-old mice, as compared to 8-week-old mice. Our results also demonstrate that tissue type, oxidative metabolic capacity and radiosensitivity influence the 3,860-bp deletion level. Therefore, this 3,860-bp deletion content may serve as a biomarker of aging and oxidative damage in mice.
Sigma-Aldrich
Ribonucleic acid, transfer from wheat germ, Type V, 15-19 units/mg solid
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), Type X-SA, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from bovine liver, Type XI, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from Escherichia coli, Type XX, Strain W, lyophilized powder