Merck
  • Repression of RNA polymerase I upon stress is caused by inhibition of RNA-dependent deacetylation of PAF53 by SIRT7.
Repression of RNA polymerase I upon stress is caused by inhibition of RNA-dependent deacetylation of PAF53 by SIRT7.
Molecular cell (2013-11-12)
Sifan Chen, Jeanette Seiler, Magaly Santiago-Reichelt, Kerstin Felbel, Ingrid Grummt, Renate Voit
ABSTRACT
Sirtuins are NAD(+)-dependent protein deacetylases that connect metabolism and cellular homeostasis. Here we show that the nuclear Sirtuin SIRT7 targets PAF53, a subunit of RNA polymerase I (Pol I). Acetylation of PAF53 at lysine 373 by CBP and deacetylation by SIRT7 modulate the association of Pol I with DNA, hypoacetylation correlating with increased rDNA occupancy of Pol I and transcription activation. SIRT7 is released from nucleoli in response to different stress conditions, leading to hyperacetylation of PAF53 and decreased Pol I transcription. Nucleolar detention requires binding of SIRT7 to nascent pre-rRNA, linking the spatial distribution of SIRT7 and deacetylation of PAF53 to ongoing transcription. The results identify a nonhistone target of SIRT7 and uncover an RNA-mediated mechanism that adapts nucleolar transcription to stress signaling.
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material