Merck
  • Regioselective synthesis of 4-substituted indoles via C-H activation: a ruthenium catalyzed novel directing group strategy.
Regioselective synthesis of 4-substituted indoles via C-H activation: a ruthenium catalyzed novel directing group strategy.
Organic letters (2013-11-28)
Veeranjaneyulu Lanke, Kandikere Ramaiah Prabhu
ABSTRACT
A highly regioselective functionalization of indole at the C-4 position by employing an aldehyde functional group as a directing group, and Ru as a catalyst, under mild reaction conditions (open flask) has been uncovered. This strategy to synthesize 4-substituted indoles is important, as this class of privileged molecules serves as a precursor for ergot alkaloids and related heterocyclic compounds.
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ruthenium, powder, 99.99% trace metals basis
Sigma-Aldrich
Ruthenium black