Merck
  • Effect of caries removal techniques on the bond strength of adhesives to caries-affected primary dentin in vitro.
Effect of caries removal techniques on the bond strength of adhesives to caries-affected primary dentin in vitro.
European journal of paediatric dentistry : official journal of European Academy of Paediatric Dentistry (2013-12-04)
E Yildiz, E Sirinkaraarslan, Z Yegin, M A Cebe, G Tosun
ABSTRACT
The aim of this in vitro study is to evaluate the effects of three different caries removal techniques on the microtensile bond strength of adhesive materials to caries-affected dentin. Thirty primary molar teeth were used. The teeth were randomly divided into three groups according to the caries removal technique employed: conventional steel bur (group 1); Er:YAG laser (group 2); chemomechanical method (group 3). Each group was divided into two subgroups according to bonding agents: one-step self-etch adhesive and etch-and-rinse adhesive. The teeth were restored with composite resin. Vertical sticks were obtained and subjected to tensile stress. Data were analyzed by two-way analysis of variance (ANOVA), Tukey's test and an independent samples t-test. The values for the laser groups were significantly lower than those of the bur groups for both bonding agents (p<0.05). There were no significant differences between the bur and chemomechanical groups (p > 0.05). Bur and chemomechanical techniques in primary teeth were found more successful. Similar results were found according to the adhesives used for each caries removal techniques.
Product Description
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)