Merck
  • Maintained cocaine self-administration is determined by quantal responses: implications for the measurement of antagonist potency.
Maintained cocaine self-administration is determined by quantal responses: implications for the measurement of antagonist potency.
The Journal of pharmacology and experimental therapeutics (2013-12-07)
Andrew B Norman, Michael R Tabet, Mantana K Norman, Vladimir L Tsibulsky
ABSTRACT
The change in frequency of cocaine self-administration as a function of the unit dose is widely assumed to represent a graded pharmacodynamic response. Alternatively, a pharmacological theory states that during maintained self-administration, a quantal response occurs at a minimum maintained cocaine concentration (satiety threshold). Rats self-administered cocaine at unit doses spanning an 8-fold range from 0.75 to 6 µmol/kg. Despite an approximately 7-fold difference in the interinjection intervals, there were no differences in the plasma cocaine concentration at the time of lever press across this range of unit doses, consistent with the satiety threshold representing an equiactive cocaine concentration. Because self-administration always occurs when cocaine concentrations decline back to the satiety threshold, this behavior represents a process of automatic back titration of equiactive agonist concentrations. Therefore, the lower frequency of self-administration at higher unit doses is caused by an increase in the duration of the cocaine-induced satiety response, and the graded dose-frequency relationship is due to cocaine pharmacokinetics. After the interinjection intervals at a particular unit dose were stable, rats were injected with the competitive D₁-like dopamine receptor antagonist R-(+)-7-chloro-8-hydroxy-3-methyl-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine (SCH23390; 15 nmol/kg intravenously) and the session continued. At all cocaine unit doses, SCH23390 accelerated self-administration with a concomitant increase in the calculated satiety threshold, and these equiactive cocaine concentration ratios were independent of the cocaine unit dose. Therefore, the measurement of antagonist potency requires only a single unit dose of cocaine, selected on the basis of convenience, and using multiple cocaine unit doses is redundant.
Product Description
Supelco
Cocaine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cocaine hydrochloride solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis
Sigma-Aldrich
Cocaine hydrochloride
Sigma-Aldrich
Cocaine free base