Merck
  • Different collagen types show distinct rates of increase from early to late stages of hepatitis C-related liver fibrosis.
Different collagen types show distinct rates of increase from early to late stages of hepatitis C-related liver fibrosis.
Human pathology (2013-12-11)
Wei Chen, Jonathan B Rock, Martha M Yearsley, Linda D Ferrell, Wendy L Frankel
ABSTRACT
During progression from normal liver to cirrhosis, total collagen increases nearly 10-fold with an abnormal increase in fibril-forming collagen and other extracellular matrix molecules. However, little is known regarding the changes each collagen type undergoes during fibrogenesis. We assessed the different collagen types by immunohistochemistry at various stages of hepatitis C-related liver fibrosis in core biopsies and compared changes in each with trichrome stain to better understand fibrogenesis. The possible utility in staging fibrosis was investigated. We found collagens III, IV, V, VI, vitronectin, and trichrome all showed statistically significant increases from early to late stages of fibrosis, but with temporal and quantitative differences. During the transition from early to late fibrosis, trichrome (stains primarily collagen I) and collagen IV showed the steepest increase and appear to be the most useful discriminators between early and late stages of fibrosis. Collagens V and VI have strong reactivity even in stage 1, which may be helpful in identifying early fibrosis when trichrome is weak or negative. Collagen III and vitronectin showed the most gradual increase. Interestingly, collagen V also showed increased staining in areas around inflammation/edema, which may overestimate established fibrosis as compared with trichrome.
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered