Merck
  • Use of novel oral anticoagulants for patients with atrial fibrillation: systematic review and clinical implications.
Use of novel oral anticoagulants for patients with atrial fibrillation: systematic review and clinical implications.
Heart & lung : the journal of critical care (2014-01-01)
Nancy M Albert
ABSTRACT
Atrial fibrillation (AF), a common arrhythmia, increases the risk of ischemic stroke. Stroke and bleeding scores for patients with AF can help to stratify risk and determine the need for antithrombotic therapy, for which warfarin has been the gold standard. Although highly effective, warfarin has several limitations that can lead to its underuse. Data from randomized, Phase III clinical trials of the novel oral anticoagulants, dabigatran, a direct thrombin inhibitor, and rivaroxaban and apixaban, both factor Xa inhibitors, indicate these drugs are at least noninferior to warfarin for the prevention of stroke and systemic embolism. They are easier to administer, and have an equivalent or lower risk of bleeding versus warfarin. A better understanding of the risks and benefits of the novel oral anticoagulants, and their use in clinical practice, will prepare clinicians to anticipate and address educational and clinical needs of AF patients and their families, and promote evidence-based prescription of appropriate and safe anticoagulation therapy.
Sigma-Aldrich
β-Alanine, 99%
Sigma-Aldrich
β-Alanine, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
β-Alanine, BioUltra, ≥99.0% (NT)
Supelco
Warfarin, PESTANAL®, analytical standard
Supelco
Warfarin, analytical standard
Sigma-Aldrich
β-Alanine, BioReagent, suitable for cell culture, suitable for insect cell culture