Merck
  • Effect of two biogas residues' application on copper and zinc fractionation and release in different soils.
Effect of two biogas residues' application on copper and zinc fractionation and release in different soils.
Journal of environmental sciences (China) (2014-02-14)
Miao Chen, Yanshan Cui, Fan Bai, Jiaojiao Wang
ABSTRACT
Biogas residue (BR) is widely used as a new green fertilizer in agriculture in China. However, it often contains a high concentration of heavy metals so its application should cause our concern. An incubation experiment was conducted to study the risk of pig biogas residue (PBR) and chicken biogas residue (CBR) application on Liuminying soil (LS) and Yixing soil (YS). The soils were incubated for one, three and six months with 0, 2%, 4% and 6% addition of BRs. According to BCR extraction results, the PBR and CBR applications induced an increase in the concentration of exchangeable fraction of Zn. As for the concentration of exchangeable fraction of Cu, an increase was only observed in the treatments with PBR application. The heavy metal binding intensity also showed a similar trend. With the PBR application, for the LS and YS, the highest concentrations of exchangeable Zn increased 3.6 and 9.5 times, respectively, while the exchangeable Cu was increased by 52.6% and 187.1%. Dissolved organic carbon was the limiting factor for the exchangeable Cu while the exchangeable Zn was controlled by soil pH. PBR presented more agricultural risk than CBR when used as fertilizer. Meanwhile, BRs were more adaptable to LS than YS according to the heavy metal release results.
Product Description
Copper (S, P), BCR®, certified reference material, chips
Copper, IRMM®, certified reference material, 0.1 mm foil
Copper, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Copper, shot, −3-+14 mesh, 99%
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Copper, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
Copper, powder, 99.99% trace metals basis
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 60-80 nm particle size (APS), ≥99.5% trace metals basis
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis