Merck
A new bergenin derivative from the rhizome of Ardisia gigantifolia.
Natural product research (2012-09-21)
Li-Hua Mu, Ju-Qiang Feng, Ping Liu
ABSTRACT
One new bergenin derivative, named 11-O-veratroylbergenin (1) along with five known bergenin derivatives, 11-O-(3'-O-methylgalloyl) bergenin (2), 11-O-syringylbergenin (3), 11-O-Galloylbergenin (4), 4-O-Galloylbergenin (5) and bergenin (6) were isolated from Ardisia gigantifolia Stapf. Their structures were established on the basis of spectroscopic analysis, especially by means of 1D, 2D-NMR and HR-ESIMS analyses. Compounds 2, 4 and 5 showed significant 2,2-diphenyl-1-picrylhydrazyl (DPPH) free-radical scavenging activity with EC₅₀ 9.7, 9.0 and 7.8 µmol L(-1), respectively. They showed more antioxidant activity than the positive control vitamin C (EC₅₀ = 28.3 µmol L(-1)).
Sigma-Aldrich
Bergenin, ≥95% (LC/MS-ELSD)