Merck
  • [Simultaneous determination of five constituents in eight Qingyedan species derived from Swertia plants by HPLC].
[Simultaneous determination of five constituents in eight Qingyedan species derived from Swertia plants by HPLC].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-08-16)
Yao-Li Li, Ming-Ying Shang, Chang-An Geng, Xue-Mei Zhang, Ji-Jun Chen, Shao-Qing Cai
ABSTRACT
To develop an HPLC method for simultaneous determination of swertiamarin, gentiopicroside, sweroside, mangiferin, erythrocentaurin, and to detect these five constituents in eight Qingyedans derived from Swertia mileensis, S. cincta, S. patens, S. punicea, S. delavayi, S. nervosa, S. macrosperma and S. yunnanensis. The separation was carried out on a Thermo BDS Hypersil C18 (4. 6 mm x 250 mm, 5 microm) column eluted with mobile phase of water containing 0. 1% phosphoric acid and methanol (B) in gradient program (0-10 min, 18%-20% B; 10-30 min, 20%-35% B; 30-35 min, 35%-60% B). The column temperature was 32 degrees C , and the detection wavelength was set at 250, 260, 225 nm. The flow rate was 0. 7 mL . min-1 from 0 to 30 min, and be increased to 1. 0 mL . min-1 in 35 min. The five compounds were well separated. The linear response ranges of swertiamarin, gentiopicroside, sweroside, mangiferin, erythrocentaurin were 0. 072-13. 39, 0. 1204. 518, 0. 060-5. 050, 0. 025-1. 518, and 0. 031-0. 210 microg, respectively. The mean recoveries of five compounds were 97.03% -102. 7% (RSD 1. 8% -6.2% ). There are swertiamarin, gentiopicroside and sweroside in most samples, and mangiferin in half samples. But erythrocentaurin was only detected in a few samples. The contents of five compounds were different in different samples. The contents of swertiamarin in S. mileensis, S. patens, S. yunnanensis and S. delavayi are up to 34. 47-118.05 mg . g-1, the contents of gentiopicroside are up to 25. 91 mg . g-1 in S. cincta. In S. puncea all contents of swertiamarin, gentiopicroside, sweroside and mangiferin are higher, especially the content of sweroside. There are Xiao-Qingyedans and Da-Qingyedans called in markets, and they can be identified by the contents of swertiamarin, gentiopicroside and sweroside. S. punicea can be identified by the content of sweroside, and the ratio gentiopicroside/total content can be used for identification of S. cincta from other seven Qingyedan species. The method was certified to be accurate and reliable and can be used for identification and quality evaluation of traditional Chinese medicine Qingyedan derived from Swertia species.
Product Description
Supelco
Swertiamarin, analytical standard
Sigma-Aldrich
Swertiamarin, ≥95% (LC/MS-ELSD)
Sigma-Aldrich
Sweroside, ≥95% (LC/MS-ELSD)