  • Search Results
  • C1q-tumour necrosis factor-related protein 8 (CTRP8) is a novel interaction partner of relaxin receptor RXFP1 in human brain cancer cells.
C1q-tumour necrosis factor-related protein 8 (CTRP8) is a novel interaction partner of relaxin receptor RXFP1 in human brain cancer cells.
The Journal of pathology (2013-09-10)
Aleksandra Glogowska, Usakorn Kunanuvat, Jörg Stetefeld, Trushar R Patel, Thatchawan Thanasupawat, Jerry Krcek, Ekkehard Weber, G William Wong, Marc R Del Bigio, Cuong Hoang-Vu, Sabine Hombach-Klonisch, Thomas Klonisch
ABSTRACT
We report a novel ligand-receptor system composed of the leucine-rich G-protein-coupled relaxin receptor, RXFP1, and the C1q-tumour necrosis factor-related protein 8 (CTRP8) in human primary brain cancer, a tumour entity devoid of the classical RXFP1 ligands, RLN1-3. In structural homology studies and computational docking experiments we delineated the N-terminal region of the globular C1q region of CTRP8 and the leucine-rich repeat units 7 and 8 of RXFP1 to mediate this new ligand-receptor interaction. CTRP8 secreted from HEK293T cells, recombinant human (rh) CTRP8, and short synthetic peptides derived from the C1q globular domain of human CTRP8 caused the activation of RXFP1 as determined by elevated intracellular cAMP levels and the induction of a marked pro-migratory phenotype in established glioblastoma (GB) cell lines and primary cells from GB patients. Employing a small competitor peptide, we were able to disrupt the CTRP8-RXFP1-induced increased GB motility. The CTRP8-RXFP1-mediated migration in GB cells involves the activation of PI3K and specific protein kinase C pathways and the increased production/secretion of the potent lysosomal protease cathepsin B (cathB), a known prognostic marker of GB. Specific inhibition of CTRP8-induced cathB activity effectively blocked the ability of primary GB to invade laminin matrices. Finally, co-immunoprecipitation studies revealed the direct interaction of human CTRP8 with RXFP1. Our results support a therapeutic approach in GB aimed at targeting multiple steps of the CTRP8-RXFP1 signalling pathway by a combined inhibitor and peptide-based strategy to block GB dissemination within the brain.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Cathepsin B from human placenta, lyophilized powder, ≥5 units/mg protein
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Cathepsin B from human liver, buffered aqueous solution, ≥1,500 units/mg protein (E1%/280)
Sigma-Aldrich
Cathepsin B from bovine spleen, lyophilized powder, ≥10 units/mg protein
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)