Merck
  • Secreted major Venus flytrap chitinase enables digestion of Arthropod prey.
Secreted major Venus flytrap chitinase enables digestion of Arthropod prey.
Biochimica et biophysica acta (2013-11-28)
Paulina Paszota, Maria Escalante-Perez, Line R Thomsen, Michael W Risør, Alicja Dembski, Laura Sanglas, Tania A Nielsen, Henrik Karring, Ida B Thøgersen, Rainer Hedrich, Jan J Enghild, Ines Kreuzer, Kristian W Sanggaard
ABSTRACT
Predation plays a major role in energy and nutrient flow in the biological food chain. Plant carnivory has attracted much interest since Darwin's time, but many fundamental properties of the carnivorous lifestyle are largely unexplored. In particular, the chain of events leading from prey perception to its digestive utilization remains to be elucidated. One of the first steps after the capture of animal prey, i.e. the enzymatic breakup of the insects' chitin-based shell, is reflected by considerable chitinase activity in the secreted digestive fluid in the carnivorous plant Venus flytrap. This study addresses the molecular nature, function, and regulation of the underlying enzyme, VF chitinase-I. Using mass spectrometry based de novo sequencing, VF chitinase-I was identified in the secreted fluid. As anticipated for one of the most prominent proteins in the flytrap's "green stomach" during prey digestion, transcription of VF chitinase-I is restricted to glands and enhanced by secretion-inducing stimuli. In their natural habitat, Venus flytrap is exposed to high temperatures. We expressed and purified recombinant VF chitinase-I and show that the enzyme exhibits the hallmark properties expected from an enzyme active in the hot and acidic digestive fluid of Dionaea muscipula. Structural modeling revealed a relative compact globular form of VF chitinase-I, which might contribute to its overall stability and resistance to proteolysis. These peculiar characteristics could well serve industrial purposes, especially because of the ability to hydrolyze both soluble and crystalline chitin substrates including the commercially important cleavage of α-chitin.
Sigma-Aldrich
Chitinase from Streptomyces griseus, lyophilized powder (essentially salt free), ≥200 units/g solid
Sigma-Aldrich
Chitinase from Trichoderma viride, lyophilized powder, ≥600 units/g solid