Merck
  • Specific carotenoid intake is inversely associated with the risk of breast cancer among Chinese women.
Specific carotenoid intake is inversely associated with the risk of breast cancer among Chinese women.
The British journal of nutrition (2014-02-08)
Lian Wang, Bin Li, Mei-Xia Pan, Xiong-Fei Mo, Yu-Ming Chen, Cai-Xia Zhang
ABSTRACT
The protective effect of dietary carotenoid intake on the risk of breast cancer is inconclusive. Moreover, data on dietary carotenoids in relation to breast cancer in non-Western populations are scarce. The aim of the present study was to examine the association between dietary carotenoid intake and the risk of breast cancer among Chinese women. A total of 561 cases and 561 controls who were frequency matched by age (5-year interval) and residence were recruited in the present case-control study. Dietary intake information was collected by a face-to-face interview using a validated FFQ. The OR and 95 % CI were assessed by multivariate logistic regression after adjusting for various potential confounders. An inverse association was observed between the consumption of α-carotene, β-carotene, β-cryptoxanthin and lutein/zeaxanthin and the risk of breast cancer. The multivariate-adjusted OR for the highest quartile of intake compared with the lowest quartile of intake were 0·61 (95 % CI 0·43, 0·88) for α-carotene, 0·54 (95 % CI 0·38, 0·78) for β-carotene, 0·38 (95 % CI 0·26, 0·52) for β-cryptoxanthin and 0·49 (95 % CI 0·34, 0·71) for lutein/zeaxanthin. Lycopene intake was not found to be associated with the risk of breast cancer, with the adjusted OR of 0·89 (95 % CI 0·61, 1·30). These inverse associations were more evident among pre-menopausal women and women who were exposed to second-hand smoke. The protective effect of specific carotenoid intake was observed for all subtypes of hormone receptor status of breast cancer. The present study indicated that a greater intake of specific carotenoids was associated with a decreased risk of breast cancer among Chinese women residing in Guangdong.
Sigma-Aldrich
β-Carotene, synthetic, ≥95% (HPLC), crystalline
Sigma-Aldrich
Xanthophyll, from marigold
Sigma-Aldrich
β-Carotene, synthetic, ≥93% (UV), powder
Supelco
β-Carotene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-Cryptoxanthin, ≥97% (TLC)
Supelco
α-Carotene, analytical standard
Supelco
Lutein, analytical standard
Supelco
Zeaxanthin, analytical standard