  • Search Results
  • Temporal trends (1999-2010) of perfluoroalkyl acids in commonly consumed food items.
Temporal trends (1999-2010) of perfluoroalkyl acids in commonly consumed food items.
Environmental pollution (Barking, Essex : 1987) (2014-03-04)
Jana H Johansson, Urs Berger, Robin Vestergren, Ian T Cousins, Anders Bignert, Anders Glynn, Per Ola Darnerud
ABSTRACT
The aim of this study was to determine how dietary exposure to PFAAs has changed over the period when major production changes occurred. Archived samples (1999-2010) of eggs, milk and farmed rainbow trout were analyzed by ultra performance liquid chromatography coupled to tandem mass spectrometry. Statistically significant decreasing trends were observed for concentrations of perfluorooctane sulfonic acid (PFOS) and perfluorohexane sulfonic acid (PFHxS) in fish (p < 0.002 and p < 0.032, respectively) and eggs (p < 0.001 for both compounds). Concentrations of PFOS in fish and eggs decreased by a factor of 10 and 40, respectively. In eggs there was also a statistically significant decreasing trend in concentrations of perfluorooctanoic acid (PFOA). The results of this study demonstrate that PFAA concentrations in food items from agricultural food chains and aquatic food chains close to sources respond rapidly to changes in environmental emissions. Implications for the overall understanding of human exposure are discussed.
Sigma-Aldrich
Pentadecafluorooctanoic acid ammonium salt, ≥98.0% (NT)
Sigma-Aldrich
Heptadecafluorooctanesulfonic acid solution, ~40% in H2O (T)
Supelco
Heptadecafluorooctanesulfonic acid solution, 100 μg/mL in methanol, analytical standard
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Sigma-Aldrich
Tetradecafluorohexane, A mixture of perfluorinated hexanes, 95%
Sigma-Aldrich
Tetradecafluorohexane, 99%
Supelco
Pentadecafluorooctanoic acid, analytical standard
Supelco
Pentadecafluorooctanoic acid solution, 100 μg/mL in methanol, analytical standard