Merck
Methemoglobinemia associated with metaflumizone poisoning.
Clinical toxicology (Philadelphia, Pa.) (2014-03-22)
J S Oh, K H Choi
ABSTRACT
Metaflumizone is a voltage-dependent sodium channel blocker insecticide, which is chemically similar to indoxacarb. Although indoxacarb poisoning is known as a cause of methemoglobinemia, the effect of metaflumizone poisoning in humans is still unknown. A 57-year-old man presented with a decreased mentality following ingestion of 100 ml of metaflumizone, 150 ml of glyphosate and alcohol. Although initial methemoglobin (MetHb) level was slightly higher than the normal limit, it gradually rose to reach a maximum level of 27.8%, on the 19 h after ingestion. After hemodialysis, MetHb level was reduced to 15.8%, which decreased further to the level of 6%, following methylene blue administration. Metaflumizone shares a similar chemical structure to indoxacarb, which is known to be a cause of methemoglobinemia. Physicians should be alert for the development of methemoglobinemia in symptomatic patients when facing potential pesticide poisoning such as metaflumizone poisoning.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
N-(Phosphonomethyl)glycine, 96%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
N-(Phosphonomethyl)glycine, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
SAFC
Glycine
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Glycine 1 M solution
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Supelco
Glyphosate, PESTANAL®, analytical standard
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution