Merck
  • Refluxing synthesis, photoluminescence and binding ability to deoxyribonucleic acid of water-soluble rare earth ion-doped LaF3 nanoparticles.
Refluxing synthesis, photoluminescence and binding ability to deoxyribonucleic acid of water-soluble rare earth ion-doped LaF3 nanoparticles.
Journal of nanoscience and nanotechnology (2014-04-18)
Zhenling Wang, Yi Zhang, Chunyang Li, Xinlei Zhang, Jiazhong Chang, Jianping Xie, Chengwei Li
ABSTRACT
Water-soluble rare earth ion (Ce3+, Tb3+)-doped LaF3 nanoparticles with the ability to bind to deoxyribonucleic acid (DNA) were prepared by the refluxing method in a glycerol/water mixture and characterized by X-ray diffraction (XRD), transmission electron microscopy (TEM), photoluminescence (PL) spectra, and so on. The obtained LaF3:Ce3+, LaF3:Tb3+ and LaF3:Ce3+, Tb3+ nanoparticles are well crystallized with a hexagonal structure and composed of spherical particles with an average size from 11 to 19 nm. The as-prepared samples can be dispersed into water to form a colloidal solution. Under ultraviolet (UV) light irradiation, the emission lines of Tb3+ in the co-doped LaF3:Ce3+, Tb3+ are evidently strengthened compared to those in the single-doped LaF3:Tb3+ nanoparticles, which is due to the energy transfer from Ce3+ to Tb3+ ions in the LaF3:Ce3+, Tb3+ samples. The biological experiment confirms that the water-soluble LaF3:Ce3+, Tb3+ nanoparticles can be bonded to the DNA molecules and emit visible light under UV irradiation. These luminescent nanoparticles could be used similarly to ethidium bromide (EtBr), which has been used extensively as a DNA staining reagent. The advantage that LaF3:Ce3+, Tb3+ nanoparticles have lower toxicity than EtBr makes them a potential reagent instead of EtBr in the DNA staining in biological experiments.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Lanthanum, pieces
Sigma-Aldrich
Lanthanum, powder, −40 mesh, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Lanthanum(III) fluoride, anhydrous, powder, 99.99% trace metals basis
Lanthanum, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, ACS reagent
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Deionized
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.