Merck
  • Bone tissue engineering by way of allograft revitalization: mechanistic and mechanical investigations using a porcine model.
Bone tissue engineering by way of allograft revitalization: mechanistic and mechanical investigations using a porcine model.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2014-04-20)
Christopher M Runyan, Samantha T Ali, Wendy Chen, Bennet W Calder, Aaron E Rumburg, David A Billmire, Jesse A Taylor
ABSTRACT
"Allograft revitalization" is a process in which cadaveric bone is used to generate well-vascularized living bone. We had previously found that porcine allograft hemimandibles filled with autologous adipose-derived stem cells (ASCs) and recombinant human bone morphogenetic protein-2-soaked absorbable collagen sponge (rhBMP-2/ACS) were completely replaced by vascularized bone, provided the construct had been incubated within a periosteal envelope. The present study sought to deepen our understanding of allograft revitalization by investigating the individual contributions of ASCs and rhBMP-2 in the process and the mechanical properties of the revitalized allograft. Porcine allograft hemimandible constructs were implanted bilaterally into rib periosteal envelopes in 8 pigs. To examine the contributions of ASCs and rhBMP-2, the following groups were assessed: group 1, periosteum alone; group 2, periosteum+ASCs; group 3, periosteum+rhBMP-2/ACS; and group 4, periosteum+ASCs+rhBMP-2/ACS. After 8 weeks, the allograft constructs were harvested for micro-computed tomography (CT) and histologic analyses and 3-point bending to assess the strength. On harvesting, the constructs receiving rhBMP-2/ACS had significantly greater bone shown by micro-CT than those receiving periosteum only (51,463 vs. 34,310 mm3; P = .031). The constructs receiving ASCs had increased bone compared to group 1 (periosteum only), although not significantly (P = .087). The combination of rhBMP-2/ACS with ASCs produced bone (50,399 mm3) equivalent to that of the constructs containing rhBMP-2/ACS only. The 3-point bending tests showed no differences between the 4 groups and a nonimplanted allograft or native mandible (P = .586), suggesting the absence of decreased strength of the allograft bone when revitalized. These data have shown that rhBMP-2/ACS significantly stimulates new bone formation by way of allograft revitalization and that the revitalized allograft has equivalent mechanical strength to native bone.
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture