Merck
  • Rapid development of migratory, linear, and serpiginous lesions in association with immunosuppression.
Rapid development of migratory, linear, and serpiginous lesions in association with immunosuppression.
Journal of the American Academy of Dermatology (2014-05-17)
Dominique C Pichard, Jennifer R Hensley, Esther Williams, Andrea B Apolo, Amy D Klion, John J DiGiovanna
ABSTRACT
A 78-year-old Bulgarian woman presented to the National Institutes of Health (NIH) with a diagnosis of poorly differentiated metastatic carcinoma of unknown origin. The prior month she had been seen at a hospital in Bulgaria for weight loss and a right inguinal mass. NIH pathology review confirmed a poorly differentiated carcinoma with extensive necrosis suggesting squamous cell carcinoma. She was enrolled in a treatment trial at NIH with metastatic disease invading the lungs and lymph nodes (mediastinum, abdomen, and pelvis) and a chemotherapy regimen was started of gemcitabine, carboplatin, and lenalidomide with dexamethasone as an antiemetic. The patient returned on day 8, and a rash of 2 days duration was noted. Immediately before arriving at the dermatology clinic, she developed altered mental status with aphasia and was admitted for neurologic observation. The altered mental status resolved and evaluation revealed only small-vessel ischemia. The patient was also experiencing diarrhea and was found to have elevated transaminases (4- to 7-fold over normal). Chemotherapy was held because of the transaminase abnormalities and altered mental status. The following day, the patient was seen by dermatology for a progressive asymptomatic eruption.
Product Description
Ivermectin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ivermectin