Merck
An evaluation of bicalutamide in the treatment of prostate cancer.
Expert opinion on pharmacotherapy (2002-08-21)
Paul F Schellhammer
ABSTRACT
Prostate cancer is a major health problem in men, causing significant morbidity and mortality. Although traditionally considered a disease of old age, improved diagnostic techniques have resulted in earlier diagnosis and many men are now treated while still physically and sexually active. Current therapies for prostate cancer, which include medical or surgical castration, have a significant impact on many aspects of quality of life. The non-steroidal antiandrogen bicalutamide (Casodex, AstraZeneca) has a favourable tolerability profile with demonstrated efficacy in several stages of prostate cancer and represents an alternative therapeutic strategy to castration. Mature survival data from men with previously untreated, locally-advanced disease reveal that bicalutamide monotherapy provides survival benefits that do not differ significantly from castration, while offering important advantages with respect to the maintenance of physical capacity and sexual interest. Recent data from a prospective randomised trial, the largest prostate cancer treatment study ever conducted, demonstrate that immediate therapy with bicalutamide (alone or as an adjuvant to therapy of curative intent) significantly reduces the risk of objective disease progression in patients with localised or locally-advanced prostate cancer. Antiandrogens are also used in combination with castration (combined androgen blockade) for advanced disease. Another large, randomised trial demonstrated that combined androgen blockade with bicalutamide is associated with a similar survival outcome to combined androgen blockade with flutamide and is better tolerated. The evidence reviewed demonstrates that bicalutamide currently has a favourable risk:benefit ratio in several stages of prostate cancer. The role of bicalutamide will be further defined by ongoing studies.
Product Description
Bicalutamide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bicalutamide (CDX), ≥98% (HPLC), powder
Bicalutamide, European Pharmacopoeia (EP) Reference Standard