Merck
High-potency sodium fluoride: a literature review.
Compendium of continuing education in dentistry (Jamesburg, N.J. : 1995) (1997-01-01)
E A Tavss, C Y Bontá, M T Joziak, S W Fisher, S K Campbell
ABSTRACT
A considerable number of clinical studies are documented in the literature on oral products containing 5,000 ppm fluoride (F), mainly nonabrasive gels. Study results show that this level of fluoride provides marked caries protection. These high-potency gels are especially effective for irradiated head and neck cancer patients, who typically develop rampant caries. Radiation therapy causes a marked decrease in salivation (xerostomia), which predisposes the teeth to caries formation and results in devastating consequences. The currently recommended treatment for these patients involves a 2-step regimen using a 5,000 ppm F gel and a conventional 1,000 ppm F dentifrice. Unfortunately, this treatment usually results in noncompliance because two steps are inconvenient for most patients. A 5,000 ppm F dentifrice meets the oral-hygiene needs of these patients. It provides an effective level of fluoride and a convenient one-step application procedure that increases patient compliance.
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium Fluoride Solution