Merck
A review and biological risk assessment of sodium saccharin.
Regulatory toxicology and pharmacology : RTP (1992-06-01)
C I Chappel
ABSTRACT
Dietary sodium saccharin is associated with bladder tumors when fed at high levels to the male rat. Under these conditions urinary pH, sodium concentration, and volume are elevated and proliferative changes are present in the urothelium. Extensive epidemiological studies have shown that saccharin does not increase the risk of bladder cancer in humans and laboratory investigations have shown that sodium saccharin is not mutagenic and does not bind to DNA. Recent research indicates that the urothelium in male rats is damaged under conditions of high urinary pH and sodium levels by a mechanism that involves alpha 2u-globulin and possibly silicate crystalluria. These studies and their implications for human health risk are reviewed.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 51143091, Saccharin, traceable to primary standards (LGC)
Sigma-Aldrich
Saccharin, ≥99%
Sigma-Aldrich
Saccharin, ≥98%
USP
Saccharin, United States Pharmacopeia (USP) Reference Standard
Supelco
Saccharin, Pharmaceutical Secondary Standard; Certified Reference Material
Saccharin, European Pharmacopoeia (EP) Reference Standard