Merck
[The research progress of succinic acid fermentation strains].
Sheng wu gong cheng xue bao = Chinese journal of biotechnology (2007-09-08)
Qing-Zhao Wang, Xue-Ming Zhao
ABSTRACT
The potential of succinic acid as an important chemical intermediates had been realized and fermentation is one of the best ways to make it possible in economical aspect. Fermentation organism is the key part of the fermentation method. The updated research developments of fermentation organisms and the fermentation characteristics and problems of them were reviewed and analyzed in this paper. Finally,the development future of fermenation organism was forecasted.
Product Description
Supelco
Succinic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Succinic acid, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Succinic acid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Succinic acid, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T)
Supelco
Succinic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland