Merck
  • Structure, function and biosynthesis of the Mycobacterium tuberculosis cell wall: arabinogalactan and lipoarabinomannan assembly with a view to discovering new drug targets.
Structure, function and biosynthesis of the Mycobacterium tuberculosis cell wall: arabinogalactan and lipoarabinomannan assembly with a view to discovering new drug targets.
Biochemical Society transactions (2007-10-25)
L J Alderwick, H L Birch, A K Mishra, L Eggeling, G S Besra
ABSTRACT
In spite of effective antibiotics to treat TB (tuberculosis) since the early 1960s, we enter the new millennium with TB, currently the leading cause of death from a single infectious agent, killing more than three million people worldwide each year. Thus an understanding of drug-resistance mechanisms, the immunobiology of cell wall components to elucidate host-pathogen interactions and the discovery of new drug targets are now required for the treatment of TB. Above the plasma membrane is a classical chemotype IV PG (peptidoglycan) to which is attached the macromolecular structure, mycolyl-arabinogalactan, via a unique diglycosylphosphoryl bridge. This review will discuss the assembly of the mAGP (mycolyl-arabinogalactan-peptidoglycan), its associated glycolipids and the site of action of EMB (ethambutol), bringing forward a new era in TB research and focus on new drugs to combat multidrug resistant TB.
Sigma-Aldrich
(+)-Arabinogalactan, ≥80% (HPLC), from larch wood