Merck
  • Toxicity, biomarkers, genotoxicity, and carcinogenicity of trichloroethylene and its metabolites: a review.
Toxicity, biomarkers, genotoxicity, and carcinogenicity of trichloroethylene and its metabolites: a review.
Journal of environmental science and health. Part C, Environmental carcinogenesis & ecotoxicology reviews (2009-08-07)
Shams Tabrez, Masood Ahmad
ABSTRACT
Trichloroethylene (TCE) is a prevalent occupational and environmental contaminant that has been reported to cause a variety of toxic effects. This article reviews toxicity, mutagenicity, and carcinogenicity caused by the exposure of TCE and its metabolites in the living system as well as on their (TCE and its metabolites) toxicity biomarkers.
Product Description
Supelco
Trichloroethylene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Trichloroethylene, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%
Supelco
Trichloroethylene, analytical standard, stabilized with 30 – 50 ppm Diisopropylamine
Sigma-Aldrich
Trichloroethylene, ACS reagent, ≥99.5%
USP
Residual Solvent Class 2 - Trichloroethylene, United States Pharmacopeia (USP) Reference Standard