Merck
Use of the adamantane structure in medicinal chemistry.
Current medicinal chemistry (2010-09-23)
Guy Lamoureux, Graciela Artavia
ABSTRACT
This review summarizes the importance and recent discoveries of the use of adamantane derivatives in Medicinal Chemistry. We have organized the article in 4 sections: 1) Absorption, Distribution, Metabolism, or Excretion (ADME) properties 2) Hydrophobic Effects 3) Ion Channels and 4) Rigid scaffold. Within each section, we have provided examples of how the adamantane group changes the properties of known drugs or provides a important pharmacophore for the design of new drugs.
Sigma-Aldrich
Adamantane, ≥99%