Merck
  • Development of cytarabine prodrugs and delivery systems for leukemia treatment.
Development of cytarabine prodrugs and delivery systems for leukemia treatment.
Expert opinion on drug delivery (2010-10-23)
Bhupender S Chhikara, Keykavous Parang
ABSTRACT
Cytarabine is a polar nucleoside drug used for the treatment of myeloid leukemia and non-Hodgkin's lymphoma. The drug has a short plasma half-life, low stability and limited bioavailability. Overdosing of patients with continuous infusions may lead to side effects. Thus, various prodrug strategies and delivery systems have been explored extensively to enhance the half-life, stability and delivery of cytarabine. Among the recent cytarabine prodrugs, amino acid conjugate ValCytarabine and fatty acid derivative CP-4055 (in Phase III trials) have been investigated for the treatment of leukemia and solid tumors, respectively. Alternatively, delivery systems of cytarabine have emerged for the treatment of different cancers. The liposomal-cytarabine formulation (DepoCyt®, Pacira Pharmaceuticals Inc., New Jersey, USA) has been approved for the treatment of lymphomatous meningitis. Various prodrug strategies evaluated for cytarabine are discussed. Then, the review summarizes the drug delivery systems that have been used for more effective cancer therapy. This review provides in-depth discussion of the prodrug strategy and delivery systems of cytarabine derivatives for the treatment of cancer. The design of cytarabine prodrugs and delivery systems provides insights for designing the next generation of more effective anticancer agents with enhanced delivery and stability. Strategies on designing cytarabine prodrug and delivery formulations showed great promise in developing effective anticancer agents with better therapeutic profile. Similar studies with other anticancer nucleosides can be an alternative approach to gaining access to more effective anticancer agents.
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, ≥90% (HPLC), crystalline
Cytarabine, European Pharmacopoeia (EP) Reference Standard