Merck
  • Functional impact and prevalence of polymorphisms involved in the hepatic glucuronidation of valproic acid.
Functional impact and prevalence of polymorphisms involved in the hepatic glucuronidation of valproic acid.
Pharmacogenomics (2012-07-31)
Dimitrios Chatzistefanidis, Ioannis Georgiou, Athanassios P Kyritsis, Sofia Markoula
ABSTRACT
Metabolism of valproic acid, a widely used drug, is only partially understood. It is mainly metabolized through glucuronidation and acts as a substrate for various UDP-glucuronosyltransferases (UGTs). UGTs metabolizing valproic acid in the liver are UGT1A3, UGT1A4, UGT1A6, UGT1A9 and UGT2B7, with UGT1A6 and UGT2B7 being the most prominent. Polymorphisms in genes expressing these enzymes may have clinical consequences, regarding dosing, blood levels of the drug and adverse reactions. Not all genes are well studied and studies, where they exist, report conflicting results. Prevalence of polymorphisms and various haplotypes is also of great importance, as it may suggest different therapeutic approaches in various populations. Presented here is a review of currently known polymorphisms, their functional impact, when known, and their prevalence in different populations, highlighting the current state of understanding and areas where there is a lack of data and suggesting new perspectives for further research.
Product Description
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propylpentanoic acid
USP
Valproic acid, United States Pharmacopeia (USP) Reference Standard
Valproic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®