Merck
  • Soluble microbial products from water biological treatment process: a review.
Soluble microbial products from water biological treatment process: a review.
Water environment research : a research publication of the Water Environment Federation (2014-04-17)
Jia Kang, Gang Du, Xu Gao, Bin Zhao, Jinsong Guo
ABSTRACT
The relationship between soluble microbial products (SMPs) and extracellular polymeric substances is described, and the characteristics of SMPs in the biological wastewater treatment process, including molecular weight distribution, metal-chelating property, biodegradability, biotoxicity, and membrane fouling, are investigated. The SMPs produced by autotrophs are degradable and utilizable for heterotrophs, thereby confirming the biodegradation of SMPs. Soluble microbial product models are designed through three approaches: establishment of SMP kinetic models or combination with Monod equations, incorporation of SMP generation and degradation into the unified theory raised by Laspidou and Rittmann (2002a), and introduction of the concept of SMP into activated sludge models. The effects of process parameters on SMP concentration are elaborated, based on the optimum biological treatment process operating parameters that can effectively minimize SMP production. The progress of SMP research in water biotreatment systems is presented, and suggestions for future studies are made.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Deionized
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.