Merck
Radiofrequency ablation for early-stage nonsmall cell lung cancer.
BioMed research international (2014-07-06)
Takao Hiraki, Hideo Gobara, Toshihiro Iguchi, Hiroyasu Fujiwara, Yusuke Matsui, Susumu Kanazawa
ABSTRACT
This review examines studies of radiofrequency ablation (RFA) of nonsmall cell lung cancer (NSCLC) and discusses the role of RFA in treatment of early-stage NSCLC. RFA is usually performed under local anesthesia with computed tomography guidance. RFA-associated mortality, while being rare, can result from pulmonary events. RFA causes pneumothorax in up to 63% of cases, although pneumothorax requiring chest drainage occurs in less than 15% of procedures. Other severe complications are rare. After RFA of stage I NSCLC, 31-42% of patients show local progression. The 1-, 2-, 3-, and 5-year overall survival rates after RFA of stage I NSCLC were 78% to 100%, 53% to 86%, 36% to 88%, and 25% to 61%, respectively. The median survival time ranged from 29 to 67 months. The 1-, 2-, and 3-year cancer-specific survival rates after RFA of stage I NSCLC were 89% to 100%, 92% to 93%, and 59% to 88%, respectively. RFA has a higher local failure rate than sublobar resection and stereotactic body radiation therapy (SBRT). Therefore, RFA may currently be reserved for early-stage NSCLC patients who are unfit for sublobar resection or SBRT. Various technologies are being developed to improve clinical outcomes of RFA for early-stage NSCLC.
Product Description
Supelco
Propofol-D17 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®