Merck
  • The co-chaperone carboxyl terminus of Hsp70-interacting protein (CHIP) mediates alpha-synuclein degradation decisions between proteasomal and lysosomal pathways.
The co-chaperone carboxyl terminus of Hsp70-interacting protein (CHIP) mediates alpha-synuclein degradation decisions between proteasomal and lysosomal pathways.
The Journal of biological chemistry (2005-04-23)
Youngah Shin, Jochen Klucken, Cam Patterson, Bradley T Hyman, Pamela J McLean
ABSTRACT
Alpha-synuclein is a major component of Lewy bodies, the pathological hallmark of Parkinson disease, dementia with Lewy bodies, and related disorders. Misfolding and aggregation of alpha-synuclein is thought to be a critical cofactor in the pathogenesis of certain neurodegenerative diseases. In the current study, we investigate the role of the carboxyl terminus of Hsp70-interacting protein (CHIP) in alpha-synuclein aggregation. We demonstrate that CHIP is a component of Lewy bodies in the human brain, where it colocalizes with alpha-synuclein and Hsp70. In a cell culture model, endogenous CHIP colocalizes with alpha-synuclein and Hsp70 in intracellular inclusions, and overexpression of CHIP inhibits alpha-synuclein inclusion formation and reduces alpha-synuclein protein levels. We demonstrate that CHIP can mediate alpha-synuclein degradation by two discrete mechanisms that can be dissected using deletion mutants; the tetratricopeptide repeat domain is critical for proteasomal degradation, whereas the U-box domain is sufficient to direct alpha-synuclein toward the lysosomal degradation pathway. Furthermore, alpha-synuclein, synphilin-1, and Hsp70 all coimmunoprecipitate with CHIP, raising the possibility of a direct alpha-synuclein-CHIP interaction. The fact that the tetratricopeptide repeat domain is required for the effects of CHIP on alpha-synuclein inclusion morphology, number of inclusions, and proteasomal degradation as well as the direct interaction of CHIP with Hsp70 implicates a cooperation of CHIP and Hsp70 in these processes. Taken together, these data suggest that CHIP acts a molecular switch between proteasomal and lysosomal degradation pathways.