Merck
  • A possible role of chenodeoxycholic acid and glycine-conjugated bile acids in fibrotic steatohepatitis in a dietary rat model.
A possible role of chenodeoxycholic acid and glycine-conjugated bile acids in fibrotic steatohepatitis in a dietary rat model.
Digestive diseases and sciences (2014-01-23)
Xiaofang Jia, Yudai Suzuki, Hisao Naito, Husna Yetti, Kazuya Kitamori, Yumi Hayashi, Rina Kaneko, Mina Nomura, Yukio Yamori, Kei Zaitsu, Masashi Kato, Akira Ishii, Tamie Nakajima
ABSTRACT
Our previous study indicated that hepatic bile acids (BAs) may have deposited and stimulated the pathogenesis of a high fat-cholesterol (HFC) diet-induced fibrotic steatohepatitis in stroke-prone spontaneously hypertensive 5/Dmcr rats, based on dysregulated BA homeostasis pathways. We aimed to further characterize BA profiles in liver and evaluate their relationships to liver injury using this model. Hepatic 21 BA levels were determined by ultra-performance liquid chromatography-tandem mass spectrometry, and their correlations with macrovesicular steatosis score, serum alanine aminotransferase (ALT) level and quantified fibrotic area were assessed using Spearman and Pearson correlations. Compared to control, BAs highly accumulated in HFC-fed rat liver at 2 weeks: cholic acid (CA), deoxycholic acid (DCA) and chenodeoxycholic acid (CDCA) were major species, thereafter, levels of CA and DCA declined, but CDCA species persistently increased, which induced a decrease in total CA/total CDCA ratio at 8 and 14 weeks. CDCA species positively, while total CA/total CDCA negatively, correlated with macrovesicular steatosis score, serum ALT and quantified fibrotic area. Unlike control, total ursodeoxycholic acid was minor in HFC-fed rat liver, and inversely correlated to aforementioned indicators of liver injury; total glyco-BAs, rather than tauro-BAs, were predominant in HFC-fed rat liver, and positively correlated with macrovesicular steatosis score. Moreover, its ratio to total tauro-BAs positively correlated with each parameter of liver injury, while inverse associations were detected for total tauro-BAs. Hepatic BA accumulation may potentiate liver disease. CDCA and glyco-BAs play a more important role in the pathogenesis of fibrotic steatohepatitis.
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
Chenodeoxycholic acid
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Chenodeoxycholic acid, European Pharmacopoeia (EP) Reference Standard
Cholic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Ammonium sulfate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis