Merck
Structure of an agonist-bound ionotropic glutamate receptor.
Science (New York, N.Y.) (2014-08-12)
Maria V Yelshanskaya, Minfen Li, Alexander I Sobolevsky
ABSTRACT
Ionotropic glutamate receptors (iGluRs) mediate most excitatory neurotransmission in the central nervous system and function by opening their ion channel in response to binding of agonist glutamate. Here, we report a structure of a homotetrameric rat GluA2 receptor in complex with partial agonist (S)-5-nitrowillardiine. Comparison of this structure with the closed-state structure in complex with competitive antagonist ZK 200775 suggests conformational changes that occur during iGluR gating. Guided by the structures, we engineered disulfide cross-links to probe domain interactions that are important for iGluR gating events. The combination of structural information, kinetic modeling, and biochemical and electrophysiological experiments provides insight into the mechanism of iGluR gating.
Sigma-Aldrich
L-Glutamic acid hydrochloride, ≥99% (HPLC)
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
SAFC
L-Cysteine
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Supelco
L-Cysteine, certified reference material, TraceCERT®
Supelco
L-Glutamic acid, certified reference material, TraceCERT®