Merck
  • The active enhancer network operated by liganded RXR supports angiogenic activity in macrophages.
The active enhancer network operated by liganded RXR supports angiogenic activity in macrophages.
Genes & development (2014-07-18)
Bence Daniel, Gergely Nagy, Nasun Hah, Attila Horvath, Zsolt Czimmerer, Szilard Poliska, Tibor Gyuris, Jiri Keirsse, Conny Gysemans, Jo A Van Ginderachter, Balint L Balint, Ronald M Evans, Endre Barta, Laszlo Nagy
ABSTRACT
RXR signaling is predicted to have a major impact in macrophages, but neither the biological consequence nor the genomic basis of its ligand activation is known. Comprehensive genome-wide studies were carried out to map liganded RXR-mediated transcriptional changes, active binding sites, and cistromic interactions in the context of the macrophage genome architecture. The macrophage RXR cistrome has 5200 genomic binding sites, which are not impacted by ligand. Active enhancers are characterized by PU.1 binding, an increase of enhancer RNA, and P300 recruitment. Using these features, 387 liganded RXR-bound enhancers were linked to 226 genes, which predominantly reside in CTCF/cohesin-limited functional domains. These findings were molecularly validated using chromosome conformation capture (3C) and 3C combined with sequencing (3C-seq), and we show that selected long-range enhancers communicate with promoters via stable or RXR-induced loops and that some of the enhancers interact with each other, forming an interchromosomal network. A set of angiogenic genes, including Vegfa, has liganded RXR-controlled enhancers and provides the macrophage with a novel inducible program.
Sigma-Aldrich
Ribonucleic acid from baker′s yeast (S. cerevisiae)
Sigma-Aldrich
Ribonucleic acid from torula yeast, Type VI
Sigma-Aldrich
Ribonucleic acid from torula yeast, core, Type II-C
Sigma-Aldrich
Ribonucleic acid diethylaminoethanol salt, Type IX
Sigma-Aldrich
LG100268, ≥98% (HPLC)
Ribonucleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
AM580, ≥98% (HPLC)