- A novel approach to reconstruct a large full thickness abdominal wall defect: successful treatment with MatriDerm® and Split.
A novel approach to reconstruct a large full thickness abdominal wall defect: successful treatment with MatriDerm® and Split.
Journal of wound care (2014-07-22)
E Tong, F Martin, O Shelley
PMID25041311
ABSTRACT
Reconstruction of large abdominal wall defects is a challenging procedure, often contraindicated in critically ill patients, with high incidences of complications. We present a case of a patient with a large abdominal wall defect who had reconstruction with MatriDerm® (an engineered dermal template) and split thickness skin grafting (SSG) as an alternative to reconstruction with myo- or fascio-cutaneous flaps. Successful wound closure was achieved with a single technique, while providing a robust reconstruction and an interface between skin and underlying viscera, should further surgery be necessary.
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered