Merck
  • 2- and 3-substituted imidazo[1,2-a]pyrazines as inhibitors of bacterial type IV secretion.
2- and 3-substituted imidazo[1,2-a]pyrazines as inhibitors of bacterial type IV secretion.
Bioorganic & medicinal chemistry (2014-12-03)
James R Sayer, Karin Walldén, Thomas Pesnot, Frederick Campbell, Paul J Gane, Michela Simone, Hans Koss, Floris Buelens, Timothy P Boyle, David L Selwood, Gabriel Waksman, Alethea B Tabor
ABSTRACT
A novel series of 8-amino imidazo[1,2-a]pyrazine derivatives has been developed as inhibitors of the VirB11 ATPase HP0525, a key component of the bacterial type IV secretion system. A flexible synthetic route to both 2- and 3-aryl substituted regioisomers has been developed. The resulting series of imidazo[1,2-a]pyrazines has been used to probe the structure-activity relationships of these inhibitors, which show potential as antibacterial agents.