Merck
  • Gelatin hydrogel as a carrier of recombinant human fibroblast growth factor-2 during rat mandibular distraction.
Gelatin hydrogel as a carrier of recombinant human fibroblast growth factor-2 during rat mandibular distraction.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2014-05-27)
Atsushi Kimura, Yuji Kabasawa, Yasuhiko Tabata, Kazuhiro Aoki, Keiichi Ohya, Ken Omura
ABSTRACT
The aim of the present study was to evaluate the feasibility of a gelatin hydrogel system to enhance recombinant human fibroblast growth factor-2 (rhFGF-2)-induced osteogenic effects during rat mandibular distraction. Mandibular distraction was performed in 28 male Wistar rats. Then, the rats were divided into 5 groups in which the designated gel mix was inserted into the distracted area: group 1, rhFGF-2 alone (n = 5); group 2, collagen alone (n = 6); group 3, collagen incorporating rhFGF-2 (n = 6); group 4, gelatin hydrogel alone (n = 5); and group 5, gelatin hydrogel incorporating rhFGF-2 (n = 6). The mandibles were excised 29 days after surgery and the newly formed bone was analyzed radiologically and histologically. The experimental groups were compared using the Fisher post hoc test (95% statistical significance threshold; P < .05). Peripheral quantitative computed tomographic analysis, von Kossa staining, and calcein staining showed that using gelatin hydrogel with rhFGF-2 (group 5) significantly increased cortical bone mineral density, the domain area of hard tissue, the domain area of cortical bone area, total bone mineral content, cortical bone mineral content, the von Kossa-stained area, and the calcein-stained area compared with the collagen carrier (group 3). Group 5 also had a significantly larger number of cells positive for tartrate-resistant acid phosphatase compared with group 3 and radiopaque areas were observed more frequently. The present findings suggest that gelatin hydrogel is a feasible delivery system for rhFGF-2, and when used together perform better in regard to hard tissue healing and treatment time after surgery.
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Phosphatase, Acid from wheat germ, ≥0.4 unit/mg solid
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 10,000, contains MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 6,000, contains 1000 ppm 4-methoxyphenol as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 20,000, contains MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 2000, contains ~1000 ppm MeHQ as stabilizer
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 550, contains 80-120 ppm MEHQ as inhibitor, 270-330 ppm BHT as inhibitor
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder