Merck
  • Autonomous bottom-up fabrication of three-dimensional nano/microcellulose honeycomb structures, directed by bacterial nanobuilder.
Autonomous bottom-up fabrication of three-dimensional nano/microcellulose honeycomb structures, directed by bacterial nanobuilder.
Journal of bioscience and bioengineering (2014-05-07)
Tetsuo Kondo, Wakako Kasai
ABSTRACT
We investigated the autonomous bottom-up fabrication of three-dimensional honeycomb cellulose structures, using Gluconacetobacter xylinus as a bacterial nanoengine, on cellulose honeycomb templates prepared by casting water-in-oil emulsions on glass substrates (Kasai and Kondo, Macromol. Biosci., 4, 17-21, 2004). The template film had a unique molecular orientation state along the honeycomb frames, but was non-crystalline. When G. xylinus, used as a nanofiber-producing bacterium, was incubated on the honeycomb scaffold in a culture medium, it secreted cellulose nanofibers only on the upper surface of the honeycomb frame. The movement was regulated by a selective interaction between the synthesized nanofiber and the surface of the honeycomb frames of the template. The relationship between directed deposition of synthesized nanofibers and ordered fabrication from the nano- to the micro-scale could provide a novel bottom-up methodology, using bacteria, for the design of three-dimensional honeycomb structures as functional materials with nano/micro hierarchical structures, with low energy consumption.
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Cellulose, fibers, (medium)
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, acid washed, powder, suitable for column chromatography
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, powder, suitable for column chromatography
Supelco
Cellulose, acid washed, from spruce, suitable for column chromatography
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.