Merck
  • Exposure to bisphenol A from drinking canned beverages increases blood pressure: randomized crossover trial.
Exposure to bisphenol A from drinking canned beverages increases blood pressure: randomized crossover trial.
Hypertension (Dallas, Tex. : 1979) (2014-12-10)
Sanghyuk Bae, Yun-Chul Hong
ABSTRACT
Bisphenol A (BPA) is a chemical used in plastic bottles and inner coating of beverage cans, and its exposure is almost ubiquitous. BPA has been associated with hypertension and decreased heart rate variability in the previous studies. The aim of the present study was to determine whether increased BPA exposure from consumption of canned beverage actually affects blood pressure and heart rate variability. We conducted a randomized crossover trial with noninstitutionalized adults, who were aged ≥60 years and recruited from a local community center. A total of 60 participants visited the study site 3 times, and they were provided the same beverage in 2 glass bottles, 2 cans, or 1 can and 1 glass bottle at a time. The sequence of the beverage was randomized. We then measured urinary BPA concentration, blood pressure, and heart rate variability 2 hours after the consumption of each beverage. The paired t test and mixed model were used to compare the differences. The urinary BPA concentration increased after consuming canned beverages by >1600% compared with that after consuming glass bottled beverages. Systolic blood pressure adjusted for daily variance increased by ≈4.5 mm Hg after consuming 2 canned beverages compared with that after consuming 2 glass bottled beverages, and the difference was statistically significant. The parameters of the heart rate variability did not show statistically significant differences.The present study demonstrated that consuming canned beverage and consequent increase of BPA exposure increase blood pressure acutely.
Product Description
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Bisphenol A, 97%