Merck
  • Polymethylmethacrylate pulmonary embolism as a complication of percutaneous vertebroplasty in cancer patients.
Polymethylmethacrylate pulmonary embolism as a complication of percutaneous vertebroplasty in cancer patients.
The Journal of the Arkansas Medical Society (2015-02-12)
Natasa Milojkovic, Samer Homsi
ABSTRACT
Percutaneous vertebroplasty is frequently used in the treatment of vertebral body fractures due to osteoporosis, vertebral body metastasis, or myeloma. Acrylic cement of polymethylmethacrylate injected into the vertebral body can leak into the paravertebral venous system and reach the pulmonary artery via the azygos vein leading to a cement pulmonary embolism. We are presenting a case of a 78 year old woman who was found to have polymethylmethacrylate pulmonary embolism as a result of vertebroplasty used for vertebral collapse from metastatic breast cancer. The appearance of new intrapulmonary artery tubular opaque density on CXR performed post procedure is highly suggestive of the diagnosis. In this case, we are exploring the importance of clinical and radiographic correlations, as well as evaluation of the hemodynamic and perfusion effect of the cement pulmonary embolism as essential steps in the management of this condition.
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 4,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 8,000
Supelco
Poly(methyl methacrylate), analytical standard, 2,480,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 50,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 20,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 100,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 10,000
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Sigma-Aldrich
Poly(methyl methacrylate)