Merck
  • Preservatives in cosmetics in the Israeli market conform well to the EU legislation.
Preservatives in cosmetics in the Israeli market conform well to the EU legislation.
Journal of the European Academy of Dermatology and Venereology : JEADV (2014-09-02)
L Horev, M Isaksson, M Engfeldt, L Persson, A Ingber, M Bruze
ABSTRACT
Preservatives are important and frequent skin sensitizers, found in a wide range of products for personal and occupational use. According to the European legislation, some cosmetic ingredients are restricted in terms of quantity and a detailed list of ingredients must be present on the product or packaging. To examine the use of preservatives in common cosmetics on the Israeli market. Sixty different Israeli brand cosmetics, including shampoos, liquid soaps, body creams and hand creams were randomly selected. Ingredient labels were examined. The products were investigated by the chromotropic acid method for release of formaldehyde and by high performance liquid chromatography for the presence of formaldehyde, DMDM hydantoin and methylchloroisothiazolinone/methylisothiazolinone (MCI/MI) and MI content. All products but one contained a detailed list of ingredients printed on the package. According to labelling, the most prevalent preservatives in Israeli shampoos and liquid soaps were DMDM hydantoin and MCI/MI. Hand creams and body creams contained mainly parabens but also iodopropynyl butylcarbamate, phenoxyethanol and DMDM hydantoin. Formaldehyde in doses from 4 to 429 ppm, and DMDM hydantoin were detected in 38 and 16 (63% and 27%) of the products, respectively. MCI/MI was detected in 11 (18%) of the products, with highest prevalence in rinse- off products (55%). Excluding one hand cream which measured 106 ppm MI, the amount of formaldehyde, DMDM hydantoin, MCI/MI and MI was within the allowed concentrations by the European directive in all cases. In Israel, adaptation of the European directive prevails, as shown by the measurements we performed on randomly selected products.
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
2,4-Dinitrophenylhydrazine, reagent grade, 97%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Supelco
Acetonitrile, analytical standard
Supelco
Acetylacetone, analytical standard
Sigma-Aldrich
Acetylacetone, Wacker Chemie AG, ≥99.5% (GC)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Hydantoin, 98%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O