Merck
  • Serological response to 13-valent pneumococcal conjugate vaccine in children and adolescents with perinatally acquired HIV infection.
Serological response to 13-valent pneumococcal conjugate vaccine in children and adolescents with perinatally acquired HIV infection.
AIDS (London, England) (2014-09-16)
Alasdair Bamford, Peter Kelleher, Hermione Lyall, Mitch Haston, Marta Zancolli, David Goldblatt, Beate Kampmann
ABSTRACT
Children with perinatally acquired HIV (paHIV) remain at an increased risk of pneumococcal infection despite highly active antiretroviral therapy (HAART). Beyond infancy, responses to pneumococcal conjugate vaccine (PCV) remain under-investigated. There are currently no published data on serological response to 13-valent PCV (PCV13) in the HIV-infected populations. We measured pneumococcal serotype-specific IgG in 48 paHIV-infected child patients (CP), 27 young adult healthy controls (AHC) and 30 child healthy controls (CHC). Opsonophagocytic assay (OPA) titres for three PCV13-exclusive serotypes were measured in a subset of children. Serotype-specific IgG was repeated 1 and 6 months following PCV13 vaccination of CP and AHC groups. OPA titres for four serotypes were measured at the 1-month time-point. The majority of CP, CHC and AHC had serotype-specific IgG above 0.35  μg/ml at baseline, although OPA activity was undetectable for two of the three serotypes studied. Baseline IgG concentrations were significantly lower in CP than AHC for a proportion of serotypes and were strongly predictive of responses to vaccine. After adjusting for baseline, postvaccination IgG concentrations were comparable, although responses to some serotypes were impaired for CP. OPA correlated well with IgG after vaccination. Detectable HIV viral load was associated with significantly lower IgG concentration and OPA titre. Children with paHIV mount a robust serological response to PCV13 for most but not all vaccine serotypes. Viral load suppression with HAART and higher baseline IgG concentration are associated with higher postvaccination antibody levels. This has implications for HAART treatment and vaccination practices.
Sigma-Aldrich
Aluminum phosphate, 99.99% trace metals basis
Sigma-Aldrich
Aluminum phosphate, reagent grade