Merck
Inositol phosphates induce DAPI fluorescence shift.
The Biochemical journal (2014-03-29)
Bernadett Kolozsvari, Federica Parisi, Adolfo Saiardi
ABSTRACT
The polymer inorganic polyP (polyphosphate) and inositol phosphates, such as IP6 (inositol hexakisphosphate; also known as phytic acid), share many biophysical features. These similarities must be attributed to the phosphate groups present in these molecules. Given the ability of polyP to modify the excitation-emission spectra of DAPI we decided to investigate whether inositol phosphates possess the same property. We discovered that DAPI-IP6 complexes emit at approximately 550 nm when excited with light of wavelength 410-420 nm. IP5 (inositol pentakisphosphate) is also able to induce a similar shift in DAPI fluorescence. Conversely, IP3 (inositol trisphosphate) and IP4 (inositol tetrakisphosphate) are unable to shift DAPI fluorescence. We have employed this newly discovered feature of DAPI to study the enzymatic activity of the inositol polyphosphate multikinase and to monitor phytase phosphatase reactions. Finally, we used DAPI-IP6 fluorescence to determine the amount of IP6 in plant seeds. Using an IP6 standard curve this straight-forward analysis revealed that among the samples tested, borlotti beans possess the highest level of IP6 (9.4 mg/g of dry mass), whereas the Indian urad bean the lowest (3.2 mg/g of dry mass). The newly identified fluorescence properties of the DAPI-IP5 and DAPI-IP6 complexes allow the levels and enzymatic conversion of these two important messengers to be rapidly and reliably monitored.
Product Description
Supelco
Inositol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DAPI, for nucleic acid staining