Merck
  • Fap2 of Fusobacterium nucleatum is a galactose-inhibitable adhesin involved in coaggregation, cell adhesion, and preterm birth.
Fap2 of Fusobacterium nucleatum is a galactose-inhibitable adhesin involved in coaggregation, cell adhesion, and preterm birth.
Infection and immunity (2015-01-07)
S Coppenhagen-Glazer, A Sol, J Abed, R Naor, X Zhang, Y W Han, G Bachrach
ABSTRACT
Fusobacterium nucleatum is a common oral anaerobe involved in periodontitis that is known to translocate and cause intrauterine infections. In the oral environment, F. nucleatum adheres to a large diversity of species, facilitating their colonization and creating biological bridges that stabilize the multispecies dental biofilm. Many of these interactions (called coadherences or coaggregations) are galactose sensitive. Galactose-sensitive interactions are also involved in the binding of F. nucleatum to host cells. Hemagglutination of some F. nucleatum strains is also galactose sensitive, suggesting that a single galactose-sensitive adhesin might mediate the interaction of fusobacteria with many partners and targets. In order to identify the fusobacterial galactose-sensitive adhesin, a system for transposon mutagenesis in fusobacteria was created. The mutant library was screened for hemagglutination deficiency, and three clones were isolated. All three clones were found to harbor the transposon in the gene coding for the Fap2 outer membrane autotransporter. The three fap2 mutants failed to show galactose-inhibitable coaggregation with Porphyromonas gingivalis and were defective in cell binding. A fap2 mutant also showed a 2-log reduction in murine placental colonization compared to that of the wild type. Our results suggest that Fap2 is a galactose-sensitive hemagglutinin and adhesin that is likely to play a role in the virulence of fusobacteria.
Product Description
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Galactose, European Pharmacopoeia (EP) Reference Standard
Supelco
Thiamphenicol, analytical standard
USP
Galactose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Thiamphenicol
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Thiamphenicol, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Chloramphenicol, VETRANAL®, analytical standard
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
SAFC
L-Arginine
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard