Merck
  • Effects of resveratrol and irradiation upon oral squamous cell carcinoma cells.
Effects of resveratrol and irradiation upon oral squamous cell carcinoma cells.
Acta odontologica Scandinavica (2014-01-28)
Antonio Navarro Atienzar, Fabio Camacho-Alonso, Pia Lopez-Jornet
ABSTRACT
To evaluate the effects of resveratrol and irradiation on oral squamous cell carcinoma (OSCC). Resveratrol was administered at doses of 5, 10, 25, 50 and 100 µM to PE/CA-PJ15 (OSCC) cultures irradiated with different doses (1, 2.5 and 5 Gy). Effects upon cell viability, apoptosis and migration were evaluated after 24, 48 and 72 h incubation. After 72 h of incubation, the 100 µM dose of resveratrol induced the greatest decrease in cell viability at all irradiation doses. After 24, 48 and 72 h of incubation, 100 µM of resveratrol induced the greatest cell apoptosis at all irradiation doses. The greatest alterations in the distribution of the G0-G1, G2-M and S phases of the cell cycle were recorded with 50 and 100 µM of resveratrol; after 24, 48 and 72 h of incubation, both these doses resulted in an increase in the S phase, at the expense of the G0-G1 and G2-M phases. Resveratrol increases cytotoxic activity in the PE/CA-PJ15 cell line and reduces cell migration capacity, while the combination of resveratrol and irradiation exerts a synergic effect.
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Resveratrol, analytical standard
Resveratrol, European Pharmacopoeia (EP) Reference Standard