Merck
  • Plasma metabolite profiling and chemometric analyses of lung cancer along with three controls through gas chromatography-mass spectrometry.
Plasma metabolite profiling and chemometric analyses of lung cancer along with three controls through gas chromatography-mass spectrometry.
Scientific reports (2015-02-26)
Syed Ghulam Musharraf, Shumaila Mazhar, Muhammad Iqbal Choudhary, Nadeem Rizi, Atta-ur-Rahman
ABSTRACT
Lung cancer has been the most common death causing cancer in the world for several decades. This study is focused on the metabolite profiling of plasma from lung cancer (LC) patients with three control groups including healthy non-smoker (NS), smokers (S) and chronic obstructive pulmonary disease patients (COPD) samples using gas chromatography-mass spectrometry (GC-MS) in order to identify the comparative and distinguishing metabolite pattern for lung cancer. Metabolites obtained were identified through National Institute of Standards and Technology (NIST) mass spectral (Wiley registry) and Fiehn Retention Time Lock (RTL) libraries. Mass Profiler Professional (MPP) Software was used for the alignment and for all the statistical analysis. 32 out of 1,877 aligned metabolites were significantly distinguished among three controls and lung cancer using p-value ≤ 0.001. Partial Least Square Discriminant Analysis (PLSDA) model was generated using statistically significant metabolites which on external validation provide high sensitivity (100%) and specificity (78.6%). Elevated level of fatty acids, glucose and acids were observed in lung cancer in comparison with control groups apparently due to enhanced glycolysis, gluconeogenesis, lipogenesis and acidosis, indicating the metabolic signature for lung cancer.
Sigma-Aldrich
Acetic anhydride
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Myristic-d27 acid, 98 atom % D, 99% (CP)
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Acetic anhydride, 99.5%
Sigma-Aldrich
Trifluoroacetamide, 97%
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Supelco
Acetic anhydride, derivatization grade (GC derivatization), LiChropur, ≥99.0%
Supelco
Hexane, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)