Merck
  • Development of papain containing pellets produced by extrusion-spheronization: an operational stage approach.
Development of papain containing pellets produced by extrusion-spheronization: an operational stage approach.
Drug development and industrial pharmacy (2014-01-15)
Gustavo H C Varca, Patricia S Lopes, Humberto G Ferraz
ABSTRACT
The performance of the standardized extrusion-spheronization technique, operational conditions, formulation parameters and storage of the final product over the bioactivity of papain containing pellets has been evaluated to obtain an insight into the potential of the technique for the manufacture of solid protein formulations. The pellets produced were assayed in terms of biological activity - monitored at each operational stage using N-benzoyl-dl-arginine ρ-nitroanilide as a substrate, and according to the physical properties - evaluated by means of size distribution, apparent density and friability. The produced pellets presented adequate physical and mechanical properties. Monitoring biological activity at each production stage revealed that the most critical steps corresponded to drying and storage, with bioactivity decay ranging from 5 to 30% and 5 to 20% for each process. Dry mixing and extrusion did not hold any influence over papain activity, while wet massing decreased the bioactivity by approximately 0-5% and the spheronization 0-2%. The results varied as a function of the experimental conditions and formulation components. In conclusion, the extrusion--spheronization technique was suitable to produce solid multiparticulate dosage forms for papain, considering the possibility to originate pellets with relatively low bioactivity decay. However, weak points of the technique corresponded to the wet massing and drying stages as well as storage.
Product Description
Supelco
L-Cysteine hydrochloride monohydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
L-Cysteine hydrochloride hydrate, 99%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, Wacker Chemie AG, 98.5-101.0%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, BioUltra, ≥99.0% (RT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium metabisulfite, tested according to Ph. Eur.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture