Merck
  • Duration of gestation and mode of delivery affect the genes of transepithelial sodium transport in pulmonary adaptation.
Duration of gestation and mode of delivery affect the genes of transepithelial sodium transport in pulmonary adaptation.
Neonatology (2014-10-11)
Cecilia Janér, Olli M Pitkänen, Liina Süvari, Ursula Turpeinen, Anniina Palojärvi, Sture Andersson, Otto Helve
ABSTRACT
Respiratory distress due to inadequate lung liquid clearance is a significant problem in infants delivered late preterm or early term, especially by elective cesarean delivery (CD). Lung liquid clearance depends on epithelial ion transport and in animals is induced by glucocorticoids. In newborn late preterm and term infants to study airway epithelial gene expressions of epithelial sodium channel (ENaC), and the serum and glucocorticoid-inducible kinase 1 (SGK1), and their association with cortisol, mode of delivery, and gestational age (GA). Infants were delivered at 35(0/7)-41(6/7) weeks. Cortisol in umbilical cord plasma was analyzed with liquid chromatography-tandem mass spectrometry. ENaC and SGK1 mRNAs in airway epithelial cells obtained within 3 h and at 1 day postnatally were quantified with real-time PCR. ENaC and SGK1 mRNAs were significantly lower in late preterm and early term infants than in those ≥ 39(0/7) weeks. Significant correlations existed between both ENaC and SGK1 and cord cortisol and GA. In term infants, SGK1 mRNA at 1.5 h was higher after vaginal delivery than elective CD. In late preterm and early term infants, low expression of ENaC and SGK1 may parallel insufficient lung liquid clearance predisposing to respiratory distress. Lower SGK1 expression after term CD could translate into insufficient sodium and lung liquid absorption. The findings demonstrate a central role for cortisol in regulation of ENaC and potentially perinatal sodium and lung liquid clearance.
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material