Merck
  • 1-Aminobenzotriazole modulates oral drug pharmacokinetics through cytochrome P450 inhibition and delay of gastric emptying in rats.
1-Aminobenzotriazole modulates oral drug pharmacokinetics through cytochrome P450 inhibition and delay of gastric emptying in rats.
Drug metabolism and disposition: the biological fate of chemicals (2014-04-15)
Rowan A Stringer, Eckhard Weber, Bruno Tigani, Paul Lavan, Stephen Medhurst, Bindi Sohal
ABSTRACT
The simultaneous effects of the cytochrome P450 inhibitor 1-aminobenzotriazole (ABT) on inhibition of in vivo metabolism and gastric emptying were evaluated with the test compound 7-(3,5-dimethyl-1H-1,2,4-triazol-1-yl)-3-(4-methoxy-2-methylphenyl)-2,6-dimethylpyrazolo[5,1-b]oxazole(NVS-CRF38), a novel corticotropin releasing factor receptor 1 (CRF1) antagonist with low water solubility, and the reference compound midazolam with high water solubility in rats. Pretreatment of rats with 100 mg/kg oral ABT administered 2 hours before a semisolid caloric test meal markedly delayed gastric emptying. ABT increased stomach weights by 2-fold; this is likely attributable to a prosecretory effect because stomach concentrations of bilirubin were comparable in ABT and control groups. ABT administration decreased the initial systemic exposure of orally administered NVS-CRF38 and increased Tmax 40-fold, suggesting gastric retention and delayed oral absorption. ABT increased the initial systemic exposure of midazolam, however for orally (but not subcutaneously) administered midazolam, extensive variability in plasma-concentration time profiles was apparent. Careful selection of administration routes is recommended for ABT use in vivo, variable oral absorption of coadministered compounds can be expected due to a disturbance of gastrointestinal transit.
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder