Merck
  • The chronic lymphocytic leukemia clone disrupts the bone marrow microenvironment.
The chronic lymphocytic leukemia clone disrupts the bone marrow microenvironment.
Stem cells and development (2014-07-24)
Alexandre Janel, Frédérique Dubois-Galopin, Céline Bourgne, Juliette Berger, Karin Tarte, Nathalie Boiret-Dupré, Stéphane Boisgard, Pierre Verrelle, Pierre Déchelotte, Olivier Tournilhac, Marc G Berger
ABSTRACT
The systematic localization of chronic lymphocytic leukemia (CLL) B-cells in the bone marrow (BM), together with the ex vivo protective effect of stromal cells on their spontaneous apoptosis, both indicate a specific role of the BM microenvironment. In vivo, the impact of CLL cells on mesenchymal stromal cells (MSCs) remains a source of debate. Here, we quantified and expanded colony forming unit-fibroblasts (CFU-Fs) from CLL-BM under standard conditions, analyzed the expression of selected genes, and studied secretion profiles. We observed failing of CLL-BM cultures in standard conditions (45.5% vs. <0.1%), and even after adding basic fibroblast growth factor (bFGF), there were fewer CFU-F than from normal BM (1.3 vs. 40/10(6) cells respectively; P<0.01). Furthermore, their polygonal aspect and low proliferative capacity, together with the expression of 384 selected genes and a secreted set of molecules related to senescence-associated secretory phenotype indicated a state of senescence, further confirmed by the higher proportion of senescence-associated β-galactosidase (SA-βGAL)-positive cells and p16INK4a overexpression. In our hands, hypoxic conditions (5% O2) did not rescue CFU-Fs. Given the role of MSC in BM tissue organization, we studied hematons that are generally considered to be elementary BM units. These structures were rare or had even disappeared completely. When hematons were present, we systematically observed nodular B-CLL cell invasion only. These data confirm that the B-CLL clone has a marked impact on MSC and disrupts BM organization in vivo, raising new questions about in vivo pathophysiology.
Product Description
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)